Learn More
Medchemexpress LLC CramP-18 (mouse) (acetate) | 99.3% | 2567.97 | 1 MG
SDP
Catalog No. 5000398424
Encompass_Preferred
This e-commerce listing features an antibiotic peptide designed for research applications. This peptide, a CRAMP-18 analog, exhibits potent antibiotic activity against various bacterial, fungal, and tumor cells. Notably, it demonstrates this efficacy without inducing hemolytic activity, making it a valuable tool for investigations into antimicrobial and anti-cancer strategies. The product is provided in a high purity formulation, ensuring reliable experimental results.
  • Potent antibiotic activity against Gram-negative bacteria, including *S. typhimurium* and *P. aeruginosa*.
  • Exhibits antifungal activity.
  • Shows potential antitumor properties.
  • No hemolytic activity, offering a favorable safety profile for *in vitro* and *in vivo* studies.
  • High purity (99.33%) ensures experimental consistency and reliability.
  • Available in multiple quantities to suit various research needs.
Catalog No. 50-003-98424 Supplier Medchemexpress LLC Supplier No. HYP5016A1MG
This e-commerce listing features an antibiotic peptide designed for research applications. This peptide, a CRAMP-18 analog, exhibits potent antibiotic activity against various bacterial, fungal, and tumor cells. Notably, it demonstrates this efficacy without inducing hemolytic activity, making it a valuable tool for investigations into antimicrobial and anti-cancer strategies. The product is provided in a high purity formulation, ensuring reliable experimental results.
  • Potent antibiotic activity against Gram-negative bacteria, including *S. typhimurium* and *P. aeruginosa*.
  • Exhibits antifungal activity.
  • Shows potential antitumor properties.
  • No hemolytic activity, offering a favorable safety profile for *in vitro* and *in vivo* studies.
  • High purity (99.33%) ensures experimental consistency and reliability.
  • Available in multiple quantities to suit various research needs.