Learn More
Medchemexpress LLC CRAMP-18 (mouse) acetate | 99.3% | 2567.97 | 5 MG
SDP
Catalog No. 5000399938
Encompass_Preferred
CRAMP-18 (mouse) acetate is an antibiotic peptide without hemolytic activity. It has good inhibitory activity against Gram-negative bacteria such as *S. typhimurium* and *P. aeruginosa*. This peptide has potential for studying antifungal, antibacterial, and antitumor properties.
  • Antibiotic peptide
  • No hemolytic activity
  • Inhibitory activity against Gram-negative bacteria
  • Potential for studying antifungal properties
  • Potential for studying antibacterial properties
  • Potential for studying antitumor properties
Catalog No. 50-003-99938 Supplier Medchemexpress LLC Supplier No. HYP5016A5MG
CRAMP-18 (mouse) acetate is an antibiotic peptide without hemolytic activity. It has good inhibitory activity against Gram-negative bacteria such as *S. typhimurium* and *P. aeruginosa*. This peptide has potential for studying antifungal, antibacterial, and antitumor properties.
  • Antibiotic peptide
  • No hemolytic activity
  • Inhibitory activity against Gram-negative bacteria
  • Potential for studying antifungal properties
  • Potential for studying antibacterial properties
  • Potential for studying antitumor properties