Learn More
Bachem CRAMP (mouse) Trifluoroacetate
SDP
Supplier:  Bachem 4056438.0001
Encompass_Preferred
CRAMP (mouse) Trifluoroacetate, H-Gly-Leu-Leu-Arg-Lys-Gly-Gly-Glu-Lys-Ile-Gly-Glu-Lys-Leu-Lys-Lys-Ile-Gly-Gln-Lys-Ile-Lys-Asn-Phe-Phe-Gln-Lys-Leu-Val-Pro-Gln-Pro-Glu-Gln-OH, 376364-36-2, C178H302N50O46, Mr 3878.67, 1mg. CRAMP is a cathelin-related antimicrobial peptide expressed in the embryonic and adult mouse. Functional studies showed CRAMP to be a potent antibiotic against Gram-negative bacteria by inhibiting growth of a variety of bacterial strains and by permeabilizing the inner membrane of E.coli directly.
Catalog No. 50-260-095