Learn More
MSC Karcher 260 kg Crane/Forklift Emptying Device, Industrial Vacuum, 1 ea
Catalog No. 502818932
Encompass
This device is engineered for the efficient emptying of industrial vacuum containers, utilizing either a crane or a forklift truck in conjunction with a crank mechanism for controlled discharge.
  • Designed for IV and IVC containers
  • Compatible with crane or forklift operation
  • Features a maximum capacity of 260 kg
  • Utilizes a crank mechanism for controlled emptying
  • Optimized for use with industrial vacuum systems
Catalog No. 50-281-8932 Supplier MSC Supplier No. 16617201
Add to Cart
Add to Cart
Request a Quote
This device is engineered for the efficient emptying of industrial vacuum containers, utilizing either a crane or a forklift truck in conjunction with a crank mechanism for controlled discharge.
  • Designed for IV and IVC containers
  • Compatible with crane or forklift operation
  • Features a maximum capacity of 260 kg
  • Utilizes a crank mechanism for controlled emptying
  • Optimized for use with industrial vacuum systems