Learn More
Grainger Adam Equipment SHS 300A Crane Scale, 300 lb Capacity, Polycarbonate, Remote Control, 1 ea
Catalog No. 502794732
Encompass_Preferred
This industrial crane scale features durable polycarbonate housing and a large, 360-degree rotating chrome-plated steel hook and shackle with a safety catch to safely weigh suspended loads. It includes a digital LED display with 15 mm (0.6 in.) high digits for clear readability and a wireless remote control for operation from a safe distance.
  • Includes wireless remote control, rechargeable battery, charger, and two AAA batteries for the remote
  • Supports multiple weighing units: kg, lb, and Newtons
  • Features external calibration, hold function, and zero-tracking
  • Equipped with an automatic power-off function and low-battery indicator
  • Provides up to 20 hours of battery life on a full charge
  • Stabilization time is approximately 2 seconds
Catalog No. 50-279-4732 Supplier Grainger Supplier No. 19YN64
This industrial crane scale features durable polycarbonate housing and a large, 360-degree rotating chrome-plated steel hook and shackle with a safety catch to safely weigh suspended loads. It includes a digital LED display with 15 mm (0.6 in.) high digits for clear readability and a wireless remote control for operation from a safe distance.
  • Includes wireless remote control, rechargeable battery, charger, and two AAA batteries for the remote
  • Supports multiple weighing units: kg, lb, and Newtons
  • Features external calibration, hold function, and zero-tracking
  • Equipped with an automatic power-off function and low-battery indicator
  • Provides up to 20 hours of battery life on a full charge
  • Stabilization time is approximately 2 seconds