Learn More
Grainger GEMTOR Crash Axe, Drop Forged Steel, 15 in., 20,000 Volts Insulated, 1 ea
Catalog No. 502796325
Encompass_Preferred
This drop forged steel crash axe is designed for heavy-duty emergency use. It features a durable, serrated cutting edge and a rubber-coated handle insulated for protection up to 20,000 volts, ensuring reliable performance in demanding environments.
  • Drop forged steel construction for durability
  • Serrated cutting edge for effective performance
  • Rubber handle insulated for 20,000 volts
  • Non-slip textured grip
  • Overall length of 15 in.
  • Limited lifetime warranty
Catalog No. 50-279-6325 Supplier Grainger Supplier No. 19TJ63
This drop forged steel crash axe is designed for heavy-duty emergency use. It features a durable, serrated cutting edge and a rubber-coated handle insulated for protection up to 20,000 volts, ensuring reliable performance in demanding environments.
  • Drop forged steel construction for durability
  • Serrated cutting edge for effective performance
  • Rubber handle insulated for 20,000 volts
  • Non-slip textured grip
  • Overall length of 15 in.
  • Limited lifetime warranty