Learn More
Cayman Chemical CrAssphage qPCR Assay, 1 ea
SDP
Catalog No. 502943105
Encompass_Preferred
A mix of primers and probe targeting CrAssphage genomic DNA (the gene of a hypothetical protein, ID: YP_009052522.3) is provided in a tube (a 20X concentrated working solution). This is a single assay that will detect CrAssphage DNA in wastewater. Organism Type: Virus dsDNA circular Enzyme Master Mixes (recommended): Empirical qPCR mastermix, Standard qPCR enzyme mastermix Panels: Control, Wastewater Colors: HEX (CrAssphage)
Catalog No. 50-294-3105 Supplier Cayman Chemical Supplier No. 406801 EA
A mix of primers and probe targeting CrAssphage genomic DNA (the gene of a hypothetical protein, ID: YP_009052522.3) is provided in a tube (a 20X concentrated working solution). This is a single assay that will detect CrAssphage DNA in wastewater. Organism Type: Virus dsDNA circular Enzyme Master Mixes (recommended): Empirical qPCR mastermix, Standard qPCR enzyme mastermix Panels: Control, Wastewater Colors: HEX (CrAssphage)