Learn More
Ampco Crate Opener
SDP
Catalog No. 19173576 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
9 in.
Catalog No. Length (English)
19-173-576 9 in.
1 options
Catalog No. 19-173-576 Supplier Ampco Supplier No. CJ1ST
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistance crate opener. Made in the USA.
  • Forked-end crowbar with a t-top end used for hammering.
  • The forked end gives leverage when opening crates.
  • Independently tested and found to pose no risk in MRI environments using a static magnetic field of 3-Tesla and a maximum spatial gradient magnetic field of 720-Gauss/cm or less.
  • Conforms to ASTM F2503.
  • Meets the requirements established by ASTM A342.
  • Height: 2 and 3/4 in.
  • Forked end height: 1 and 3/8 in.
  • Length: 9 in.
Specifications
Product Type Crate Opener
Length (English) 9 in.
Quantity 1 Ea.