Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cratoxylone | 149155-01-1 | 5 MG
SDP

Catalog No. 5000418571
Encompass_Preferred

Cratoxylone is a compound isolated from the bark of Cratoxylum Cochinchinense, known for its antiplasmodial activity. It acts as a parasite inhibitor within the anti-infection pathway, demonstrating inhibition of NO production in LPS-stimulated mouse RAW264.7 cells.

  • Possesses antiplasmodial activity
  • Inhibits NO production in LPS-stimulated RAW264.7 cells with an IC50 of 5.4 μM
  • Appears as a solid, light yellow to yellow substance
  • Has a purity of 95.0%
  • Molecular formula C24H28O7
  • Molecular weight 428.47
  • Classified as phenols, polyphenols, and xanthones
  • Derived from plants

Catalog No. 50-004-18571 Supplier Medchemexpress LLC Supplier No. HYN62515MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cratoxylone is a compound isolated from the bark of Cratoxylum Cochinchinense, known for its antiplasmodial activity. It acts as a parasite inhibitor within the anti-infection pathway, demonstrating inhibition of NO production in LPS-stimulated mouse RAW264.7 cells.

  • Possesses antiplasmodial activity
  • Inhibits NO production in LPS-stimulated RAW264.7 cells with an IC50 of 5.4 μM
  • Appears as a solid, light yellow to yellow substance
  • Has a purity of 95.0%
  • Molecular formula C24H28O7
  • Molecular weight 428.47
  • Classified as phenols, polyphenols, and xanthones
  • Derived from plants

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.