Learn More
Aviva Systems Biology Corp CRBN Antibody - N-terminal region (ARP56882_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56882_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRBN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CRBN. Alternate names: DKFZp781K0715, MGC27358, MRT2A, MRT2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62025