Learn More
Biomatik Corporation Crbn Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10920
Encompass_Preferred
Crbn Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to CRBN. The Crbn Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Cereblon Protein (1-280Aa). Applications: ELISA, WB, IHC. Target Synonyms: CRBN. Accession Number: Q96SW2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4683