Learn More
Electron Microscopy Sciences Fischione Carbon Film on Copper Tomography Grid
SDP
Catalog No. 501931248
Encompass_Preferred
This 1.5mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90 deg.
Catalog No. 50-193-1248 Supplier Electron Microscopy Sciences Supplier No. 7435703
This 1.5mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90 deg.