Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Square 150 mesh Nickel Grid
SDP
Catalog No. 501903069
Encompass_Preferred
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Ultra-thin thickness is 3 to 4 nm.
Grid: Nickel
Catalog No. 50-190-3069 Supplier Electron Microscopy Sciences Supplier No. CF150NIUL
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Ultra-thin thickness is 3 to 4 nm.
Grid: Nickel