Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on Square 200 mesh Copper Grid
SDP
Catalog No. 501903076
Encompass_Preferred
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Extra thick thickness is 20 to 30 nm.
Grid: Copper
Catalog No. 50-190-3076 Supplier Electron Microscopy Sciences Supplier No. CF200CUET
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Extra thick thickness is 20 to 30 nm.
Grid: Copper