Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Square 300 mesh Gold Grid
SDP
Catalog No. 501900147
Encompass_Preferred
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Standard thickness is 5-6 nm.
Grid: Gold
Catalog No. 50-190-0147 Supplier Electron Microscopy Sciences Supplier No. CF300AU25
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Standard thickness is 5-6 nm.
Grid: Gold