Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on Square 300 mesh Nickel Grid
SDP
Catalog No. 501903086
Encompass_Preferred
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Extra thick thickness is 20 to 30 nm.
Grid: Nickel
Catalog No. 50-190-3086 Supplier Electron Microscopy Sciences Supplier No. CF300NIET
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Extra thick thickness is 20 to 30 nm.
Grid: Nickel