Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Square 400 mesh Gold Grid
SDP
Catalog No. 501903094
Encompass_Preferred
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Ultra-thin thickness is 3 to 4 nm.
Grid: Gold
Catalog No. 50-190-3094 Supplier Electron Microscopy Sciences Supplier No. CF400AUUL
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Ultra-thin thickness is 3 to 4 nm.
Grid: Gold