Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on Gilder F1 Finder Gold Grid
SDP
Catalog No. 501903105
Encompass_Preferred
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold
Catalog No. 50-190-3105 Supplier Electron Microscopy Sciences Supplier No. CF200F1AUET
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold