Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Gilder F1 Finder Nickel Grid
SDP
Catalog No. 501903100
Encompass_Preferred
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Nickel
Catalog No. 50-190-3100 Supplier Electron Microscopy Sciences Supplier No. CF200F1NIUL
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Nickel