Learn More
Electron Microscopy Sciences Carbon Support Film 10 nm thick on Gilder F2 Finder Nickel Grid
SDP
Catalog No. 501903110
Encompass_Preferred
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Nickel
Catalog No. 50-190-3110 Supplier Electron Microscopy Sciences Supplier No. CF200F2NITH
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Nickel