Learn More
Electron Microscopy Sciences Carbon Support Film 10 nm thick on London Finder LF135 Copper Grid
SDP
Catalog No. 501903116
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper
Thickness: 10 nm
Catalog No. 50-190-3116 Supplier Electron Microscopy Sciences Supplier No. CFLF135CUTH
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper
Thickness: 10 nm