Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on London Finder LF200 Gold Grid
SDP
Catalog No. 501900797
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Gold
Thickness: 5-6 nm
Catalog No. 50-190-0797 Supplier Electron Microscopy Sciences Supplier No. CFLF200AU25
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Gold
Thickness: 5-6 nm