Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on London Finder LF200 Nickel Grid
SDP
Catalog No. 501900796
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel
Thickness: 5-6 nm
Catalog No. 50-190-0796 Supplier Electron Microscopy Sciences Supplier No. CFLF200NI50
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel
Thickness: 5-6 nm