Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on London Finder LF400 Copper Grid
SDP
Catalog No. 501900799
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper
Thickness: 5-6 nm
Catalog No. 50-190-0799 Supplier Electron Microscopy Sciences Supplier No. CFLF400CU50
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper
Thickness: 5-6 nm