Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on London Finder LF400 Copper Grid
SDP
Catalog No. 501903130
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper
Thickness: 3-4 nm
Catalog No. 50-190-3130 Supplier Electron Microscopy Sciences Supplier No. CFLF400CUUL
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper
Thickness: 3-4 nm