Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on London Finder LF400 Copper Grid
SDP
Catalog No. 501903132
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Copper
Thickness: 20-30 nm
Catalog No. 50-190-3132 Supplier Electron Microscopy Sciences Supplier No. CFLF400CUET
A thin film of pure carbon deposited on the shiny side of a London Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Copper
Thickness: 20-30 nm