Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Gilder F2 Finder Nickel Grid
SDP
Catalog No. 501900790
Encompass_Preferred
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel
Catalog No. 50-190-0790 Supplier Electron Microscopy Sciences Supplier No. CF200F2NI50
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel