Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Hexagonal 100 mesh Nickel Grid
SDP
Catalog No. 501900812
Encompass_Preferred
Our high quality coated grids will make your microscopy work easier and save you time. A thin film of pure carbon deposited on one side of the grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in a grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel
Catalog No. 50-190-0812 Supplier Electron Microscopy Sciences Supplier No. CF100HNI50
Our high quality coated grids will make your microscopy work easier and save you time. A thin film of pure carbon deposited on one side of the grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in a grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel