Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Hexagonal 200 mesh Nickel Grid
SDP
Catalog No. 501903148
Encompass_Preferred
Our high quality coated grids will make your microscopy work easier and save you time. A thin film of pure carbon deposited on one side of the grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in a grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Nickel
Catalog No. 50-190-3148 Supplier Electron Microscopy Sciences Supplier No. CF200HNIUL
Our high quality coated grids will make your microscopy work easier and save you time. A thin film of pure carbon deposited on one side of the grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in a grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Nickel