Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Hexagonal 400 mesh Gold Grid
SDP
Catalog No. 501903169
Encompass_Preferred
Our high quality coated grids will make your microscopy work easier and save you time. A thin film of pure carbon deposited on one side of the grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in a grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold
Catalog No. 50-190-3169 Supplier Electron Microscopy Sciences Supplier No. CF400HAUUL
Our high quality coated grids will make your microscopy work easier and save you time. A thin film of pure carbon deposited on one side of the grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in a grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold