Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on Single 1000 µm hole Nickel Grid
SDP
Catalog No. 501903315
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Nickel
Catalog No. 50-190-3315 Supplier Electron Microscopy Sciences Supplier No. CFGA1000NIET
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Nickel