Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Single 75 µm hole Copper Grid
SDP
Catalog No. 501900894
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper
Catalog No. 50-190-0894 Supplier Electron Microscopy Sciences Supplier No. CFGA100CU25
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper