Learn More
Electron Microscopy Sciences Carbon Support Film 10 nm thick on Single 1500 µm hole Copper Grid
SDP
Catalog No. 501903317
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper
Catalog No. 50-190-3317 Supplier Electron Microscopy Sciences Supplier No. CFGA1500CUTH
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper