Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Single 150 µm hole Copper Grid
SDP
Catalog No. 501903274
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper
Catalog No. 50-190-3274 Supplier Electron Microscopy Sciences Supplier No. CFGA150CUUL
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper