Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Single 400 µm hole Nickel Grid
SDP
Catalog No. 501900912
Encompass_Preferred
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel
Catalog No. 50-190-0912 Supplier Electron Microscopy Sciences Supplier No. CFGA400NI25
A thin film of pure carbon deposited on the shiny side of the single hole grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel