Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CRBN ligand-11 | 2962967-27-5 | 99.9% | 301.11 | 250 MG
SDP

Catalog No. 5000394426
Encompass_Preferred

CRBN ligand-11 is an E3 ubiquitin ligase ligand for CFT8634 and can be used for the synthesis of PROTACs.

  • Targets Cereblon
  • Solid appearance
  • Color is white to off-white
  • Store at 4°C, protected from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Involves PROTAC pathway

Catalog No. 50-003-94426 Supplier Medchemexpress LLC Supplier No. 250MGHY169989
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CRBN ligand-11 is an E3 ubiquitin ligase ligand for CFT8634 and can be used for the synthesis of PROTACs.

  • Targets Cereblon
  • Solid appearance
  • Color is white to off-white
  • Store at 4°C, protected from light
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Involves PROTAC pathway

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.