Learn More
Medchemexpress LLC CRBN ligand-9 (compound 4d) | 55003-81-1 | MFCD30188076 | 98.0% | 303.23 g/mol | C13H9N3O6 | 250 MG
SDP
Catalog No. 5000282200
Encompass_Preferred
CRBN ligand-9 (Compound 4d) is a small-molecule cereblon-binding ligand used as a building block in targeted protein degradation research. It is supplied as a research-grade solid with documented purity and analytical data suitable for medicinal chemistry and PROTAC synthesis.
  • Used as a cereblon-binding building block for proteolysis-targeting chimera (PROTAC) design and synthesis.
  • Exhibits moderate cereblon affinity (Ki ≈ 8.9 μM) suitable for degrader development.
  • Provided with purity 98.0% and accompanying analytical documentation (COA, SDS).
  • Defined molecular properties: molecular weight 303.23 g/mol, formula C13H9N3O6.
  • Available in research-scale pack sizes for synthesis and assay work.
Catalog No. 50-002-82200 Supplier Medchemexpress LLC Supplier No. HYW342590250MG
Add to Cart
Add to Cart
CRBN ligand-9 (Compound 4d) is a small-molecule cereblon-binding ligand used as a building block in targeted protein degradation research. It is supplied as a research-grade solid with documented purity and analytical data suitable for medicinal chemistry and PROTAC synthesis.
  • Used as a cereblon-binding building block for proteolysis-targeting chimera (PROTAC) design and synthesis.
  • Exhibits moderate cereblon affinity (Ki ≈ 8.9 μM) suitable for degrader development.
  • Provided with purity 98.0% and accompanying analytical documentation (COA, SDS).
  • Defined molecular properties: molecular weight 303.23 g/mol, formula C13H9N3O6.
  • Available in research-scale pack sizes for synthesis and assay work.