Learn More
Cell Signaling Technology CRBN Matched Antibody Pair, Human, Mouse, Rat, Unconjugated, 1 Kit
SDP
Catalog No. 503132042
Encompass
The CRBN Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with custom or specialized labeling, such as biotin, fluorophores, lanthanides, or beads. This kit is compatible with platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Includes two rabbit monoclonal antibodies against CRBN, each supplied at 100 μg.
  • Features antibodies that bind to non-overlapping epitopes.
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8) and are BSA and azide free.
  • Validated for human, mouse, and rat species reactivity.
  • Intended for research use only, not for use in diagnostic procedures.
Catalog No. 50-313-2042 Supplier Cell Signaling Technology Supplier No. 38606S
The CRBN Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with custom or specialized labeling, such as biotin, fluorophores, lanthanides, or beads. This kit is compatible with platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Includes two rabbit monoclonal antibodies against CRBN, each supplied at 100 μg.
  • Features antibodies that bind to non-overlapping epitopes.
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8) and are BSA and azide free.
  • Validated for human, mouse, and rat species reactivity.
  • Intended for research use only, not for use in diagnostic procedures.