Learn More
Medchemexpress LLC CRBN modulator-1 | 2407829-65-4 | 98.0% | 324.29 | 100 MG
SDP
Catalog No. 5000350548
Encompass_Preferred
CRBN modulator-1 is a Thalidomide analog and a CRBN modulator, extracted from WO2020006262A1, compound 10. It has an IC50 of 3.5 μM and a Ki of 0.98 μM. This product is for research use only and not sold to patients.
  • Thalidomide analog and CRBN modulator
  • IC50 of 3.5 μM
  • Ki of 0.98 μM
  • For research use only
  • Solid appearance
  • Light green to green color
  • Purity of 98.0%
Catalog No. 50-003-50548 Supplier Medchemexpress LLC Supplier No. HY138040100MG
CRBN modulator-1 is a Thalidomide analog and a CRBN modulator, extracted from WO2020006262A1, compound 10. It has an IC50 of 3.5 μM and a Ki of 0.98 μM. This product is for research use only and not sold to patients.
  • Thalidomide analog and CRBN modulator
  • IC50 of 3.5 μM
  • Ki of 0.98 μM
  • For research use only
  • Solid appearance
  • Light green to green color
  • Purity of 98.0%