Learn More
Medchemexpress LLC CRBN modulator-1 | 2407829-65-4 | 98.0% | 324.29 | 25 MG
SDP
Catalog No. 5000350567
Encompass_Preferred
CRBN modulator-1 is a Thalidomide analog and a CRBN modulator, extracted from WO2020006262A1, compound 10. It has an IC₅₀ of 3.5 μM and a Kᵢ of 0.98 μM. This product is for research use only.
  • Thalidomide analog and CRBN modulator.
  • IC₅₀ of 3.5 μM.
  • Kᵢ of 0.98 μM.
  • For research use only.
Catalog No. 50-003-50567 Supplier Medchemexpress LLC Supplier No. 25MGHY138040
CRBN modulator-1 is a Thalidomide analog and a CRBN modulator, extracted from WO2020006262A1, compound 10. It has an IC₅₀ of 3.5 μM and a Kᵢ of 0.98 μM. This product is for research use only.
  • Thalidomide analog and CRBN modulator.
  • IC₅₀ of 3.5 μM.
  • Kᵢ of 0.98 μM.
  • For research use only.