Learn More
Medchemexpress LLC CRBN modulator-1 | 2407829-65-4 | 98.0% | 324.29 | 50 MG
SDP
Catalog No. 5000350857
Encompass_Preferred
CRBN modulator-1 is a thalidomide analog and a cereblon (CRBN) modulator. This compound, identified as compound 10 from WO2020006262A1, is presented as a solid with a light green to green color. It is intended for research use only.
  • Thalidomide analog
  • CRBN modulator
  • IC50 of 3.5 μM
  • Ki of 0.98 μM
  • Targets Cereblon
  • Solid appearance
  • Light green to green color
Catalog No. 50-003-50857 Supplier Medchemexpress LLC Supplier No. HY13804050MG
CRBN modulator-1 is a thalidomide analog and a cereblon (CRBN) modulator. This compound, identified as compound 10 from WO2020006262A1, is presented as a solid with a light green to green color. It is intended for research use only.
  • Thalidomide analog
  • CRBN modulator
  • IC50 of 3.5 μM
  • Ki of 0.98 μM
  • Targets Cereblon
  • Solid appearance
  • Light green to green color