Learn More
Medchemexpress LLC CRBN modulator-1 | 2407829-65-4 | MFCD33029299 | 98.0% | 324.29 g/mol | C17H12N2O5 | 5 MG
SDP
Catalog No. 5000222825
Encompass_Preferred
CRBN modulator-1 is a thalidomide analog and cereblon (CRBN) modulator described in WO2020006262A1 (compound 10). It has reported biochemical activity with IC50 = 3.5 μM and Ki = 0.98 μM and is intended for laboratory research use only.
  • Thalidomide analog that modulates cereblon (CRBN).
  • Reported biochemical activity: IC50 3.5 μM; Ki 0.98 μM.
  • High purity (98.0%) suitable for research applications.
  • Available in small pack sizes for assay development.
  • Useful for biochemical and cell-based screening studies.
Catalog No. 50-002-22825 Supplier Medchemexpress LLC Supplier No. HY1380405MG
CRBN modulator-1 is a thalidomide analog and cereblon (CRBN) modulator described in WO2020006262A1 (compound 10). It has reported biochemical activity with IC50 = 3.5 μM and Ki = 0.98 μM and is intended for laboratory research use only.
  • Thalidomide analog that modulates cereblon (CRBN).
  • Reported biochemical activity: IC50 3.5 μM; Ki 0.98 μM.
  • High purity (98.0%) suitable for research applications.
  • Available in small pack sizes for assay development.
  • Useful for biochemical and cell-based screening studies.