Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crbn modulator-1 | 2407829-65-4 | MFCD33029299 | 98.0% | C17H12N2O5 | 10MG
SDP

Catalog No. 5000258302
Encompass_Preferred

CRBN modulator-1 is a thalidomide analog that modulates cereblon (CRBN), an E3 ligase substrate receptor. Described in WO2020006262A1 (compound 10), the compound has reported biochemical activity (IC50 3.5 μM; Ki 0.98 μM) and is supplied as a solid for research use with supporting analytical documentation.

  • Acts as a cereblon modulator with IC50 3.5 μM and Ki 0.98 μM.
  • Provided as a solid powder suitable for research applications.
  • High purity, 98.0%, with supporting analytical data available.
  • Molecular formula C17H12N2O5; molecular weight 324.29 g/mol.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Documentation includes datasheet, certificate of analysis, NMR, HPLC, and MS.

Catalog No. 50-002-58302 Supplier Medchemexpress LLC Supplier No. HY13804010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CRBN modulator-1 is a thalidomide analog that modulates cereblon (CRBN), an E3 ligase substrate receptor. Described in WO2020006262A1 (compound 10), the compound has reported biochemical activity (IC50 3.5 μM; Ki 0.98 μM) and is supplied as a solid for research use with supporting analytical documentation.

  • Acts as a cereblon modulator with IC50 3.5 μM and Ki 0.98 μM.
  • Provided as a solid powder suitable for research applications.
  • High purity, 98.0%, with supporting analytical data available.
  • Molecular formula C17H12N2O5; molecular weight 324.29 g/mol.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Documentation includes datasheet, certificate of analysis, NMR, HPLC, and MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.