Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on 1500 µm Slot Grid Copper Grid
SDP
Catalog No. 501902064
Encompass_Preferred
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper
Catalog No. 50-190-2064 Supplier Electron Microscopy Sciences Supplier No. CF205CU50
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper