Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on 2X1 mm Slot Grid Gold Grid
SDP
Catalog No. 501903259
Encompass_Preferred
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold
Catalog No. 50-190-3259 Supplier Electron Microscopy Sciences Supplier No. CF2010AUUL
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold