Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on 2X1 mm Slot Grid Copper Grid
SDP
Catalog No. 501903253
Encompass_Preferred
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper
Catalog No. 50-190-3253 Supplier Electron Microscopy Sciences Supplier No. CF2010CUUL
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper