Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on 2X1 mm Slot Grid Copper Grid
SDP
Catalog No. 501903255
Encompass_Preferred
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Copper
Catalog No. 50-190-3255 Supplier Electron Microscopy Sciences Supplier No. CF2010CUET
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Copper