Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on 2 X 1 mm Slot Grid Nickel Grid
SDP
Catalog No. 501900882
Encompass_Preferred
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel
Catalog No. 50-190-0882 Supplier Electron Microscopy Sciences Supplier No. CF2010NI25
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel