Learn More
Electron Microscopy Sciences Carbon Support Film 10 nm thick on 2X1 mm Slot Grid Nickel Grid
SDP
Catalog No. 501903257
Encompass_Preferred
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Nickel
Catalog No. 50-190-3257 Supplier Electron Microscopy Sciences Supplier No. CF2010NITH
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Nickel