Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on 2X1 mm Slot Grid Nickel Grid
SDP
Catalog No. 501903258
Encompass_Preferred
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Nickel
Catalog No. 50-190-3258 Supplier Electron Microscopy Sciences Supplier No. CF2010NIET
A thin film of pure carbon deposited on one side of the slot grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Nickel